Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Autoclaves, front loading, D-Line

Autoclave, caricamento frontale, 2840 ELC-G-D, con raffreddamento rapido e riscaldamento efficiente tramite il vapore proveniente dal generatore, 28 L
Codice catalogo 481-0849
Nome del fornitore:   TUTTNAUER
Autoclaves, front loading, D-Line
Codice catalogo 481-0849
Nome del fornitore:  TUTTNAUER
Autoclaves, front loading, D-Line
Autoclaves, front loading, D-Line

Autoclave, caricamento frontale, 2840 ELC-G-D, con raffreddamento rapido e riscaldamento efficiente tramite il vapore proveniente dal generatore, 28 L

Price
SFr. 15’210.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceutical, food, medical and biotechnological industries. This advanced laboratory line is a quality range of reliable bench top (front loading) and vertical (top loading) models with chamber volumes from 28 to 160 litres that provides a single solution for the full spectrum of sterilisation needs. The autoclaves can be fully configured from general use to customer-specific applications. Units from either the Essential or Hi End ranges can be customised with various different optional features to meet more specific or demanding applications.

  • High quality AISI 316 L stainless steel chamber with mirror finished door
  • Advanced control system with easy to use three-button keypad
  • Up to 20 customisable cycle programs
  • Standard USB and Ethernet connection allows easy transfer of cycle data to a PC
  • Two flexible PT100 temperature sensors

The Essential bench top (front loading) laboratory autoclaves fit on benches or work tops. These basic stand-alone models are well known for their steam production inside the chamber by means of heating elements around the chamber. This allows the user to run basic sterilisation cycles for solid loads, liquids and waste (solid or liquid). Adding different options and accessories enables the user to achieve even more functionality from the autoclave such as quicker cooling times, dry products at the end and safe handling of hazardous loads.
Easy to clean chamber
Manual filling and draining of the demineralised water reservoir
Re-use of the demineralised water

The Hi End horizontal (front loading) laboratory autoclaves 2840 ELC-G-D model fits directly on benches or counter tops, where the 38xx and 50xx ELC-G-D models fit directly on the laboratory floor without the need for a support table. These models are designed to be connected directly to utilities and are well known for their steam production in the built-in steam generator as well as the standard fast cooling option. This allows the user to run liquid cycles with fast cooling in addition to the basic sterilisation cycles for solid loads, liquids and waste (solid or liquid). Adding different options and accessories enables the user to achieve even more functionality from the autoclave such as faster cooling times, dry products at the end and safe handling of hazardous loads.
Full utilities required (demineralised water (not for model 2840), tap water, drain connection, compressed air and electricity)
Automatic filling and draining functionality (optional for model 2840)
Automatic cooling of the exhaust to the drain 60 °C (by parameter)

 : Accessories as well as a range of factory fitted options such as units with fast/air cooling, super fast cooling, pre-/post-vacuum and models suitable for biohazardous waste sterilisation are available on request. Please contact your local Avantor sales office for further details prior to ordering as these cannot be retro-fitted.

 : Note: all 2840 and the Essential 3850 models require 230 V single phase and the Hi End 3850 and all 3870, 5050 and 5075 models require a 400 V 3 phase supply.

Specifiche
  • Capacità:
    28 L
  • Descrizione:
    Autoclave, caricamento frontale, 2840 ELC-G-D, con raffreddamento rapido e riscaldamento efficiente tramite il vapore proveniente dal generatore
  • Seq:
    006
  • Cat. No.:
    481-0849
  • Pressione max.:
    2,8 bar
  • Peso:
    50 kg
  • Ø×A interne:
    280×450 mm
  • Materiale:
    Acciaio inossidabile 316L
  • Temperatura:
    134 °C
  • Modello:
    2840 ELC-G-D
  • L×P×A est.:
    530×630×400 mm
  • Potenza:
    2200 W
Services

Informazioni sul prodotto

  • Codice catalogo
    Descrizione
    Per
    Disponibilità
    Price
  • 481-0794
    Trasporto dal fornitore al cliente, incluso disimballaggio e posizionamento a destinazione finale con l'utente finale (disponibile solo in Germania)
    Modelli 50XX
    Log in per disponibilità
    1 * 1 ST - SFr. 1’540.00

Accessories

Informazioni sul prodotto

  • Codice catalogo
    Per
    Disponibilità
    Price
  • TUTTCLE096-0028
    All Tuttnauer models
    Log in per disponibilità
    1 * 1 L - SFr. 27.40

Informazioni sul prodotto

  • Codice catalogo
    Per
    Disponibilità
    Price
  • Carta per stampante termica THE-002-0052 (5 rotoli)
    Carta per stampante termica THE-002-0052 (5 rotoli)
    481-0851
    Modelli EL-ELV e D-line
    Log in per disponibilità
    1 * 5 ST - SFr. 13.90

Informazioni sul prodotto

  • Codice catalogo
    Capacità
    Per
    Disponibilità
    Price
  • Contenitore in acciaio inox, 265L×390W mm, 21,5 L
    Contenitore in acciaio inox, 265L×390W mm, 21,5 L
    TUTTBSK411-0003
    __
    2840 autoclave
    Log in per disponibilità
    1 * 1 ST - SFr. 1’570.00

Informazioni sul prodotto

  • Codice catalogo
    Capacità
    Disponibilità
    Price
  • Ruote per autoclave 5075
    Ruote per autoclave 5075
    TUTTADD222-0220
    __
    Log in per disponibilità
    1 * 1 ST - SFr. 420.00