ICP-MS tuning solutions
ICP-MS stock tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co, 10 mg/L, 2% HNO₃, 100 mL
Nome del fornitore: AGILENT HEWLETT PACKARD
ICP-MS tuning solutions
Codice catalogo HEWL5190-0465
Nome del fornitore: AGILENT HEWLETT PACKARD
Numero CAS:
Vai a
Product Details & Documents
PA Tuning solution set includes Tuning 1 and Tuning 2 solutions.
Tuning 1: 20 ppm each of Zn, Be, Cd, As; 10 ppm each of Ni, Pb, Mg; 5 ppm each of Tl, Na, Al, U, Cu, Th, Ba, Co, Sr, V, Cr, Mn, 6Li, Sc, In, Lu, Bi; 2,5 ppm each of Y, Yb; matrix 2 to 5% HNO₃
Tuning 2: 10 ppm each of Mo, Sb, Sn, Ge, Ru, Pd; 5 ppm of Ti, Ir; matrix 10% HCl and 1% HNO₃ with trace amounts of HF.
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Concentrazione:10 mg/L
- Matrice:2% HNO₃
- Conf:100 mL
- Descrizione:ICP-MS stock tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co



