Racks for BOD bottles, WHEATON®
Rack, for 12 BOD bottles (300 ml)
Catalog # WHEAW227731
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Wire, coated.
- Good chemical resistance
- Good temperature resistance
These stackable racks transports BOD bottles conveniently and safely.
Specifications
- Descrizione:Rack, for 12 BOD bottles (300 ml)
- Seq:02