Orders LinkContactUsLinkComponent
Thermal transfer printer labels, type B-424
Printer labels, B-424, THT-19-424-1, Paper, Permanente, a base di gomma, Thermal transfer, bianco, 360X-Plus II, 76×50 mm, 1000 labels/roll
  BRDY030138
 :   Brady
Thermal transfer printer labels, type B-424
  BRDY030138
 :  Brady
Thermal transfer printer labels, type B-424
Thermal transfer printer labels, type B-424
Printer labels, B-424, THT-19-424-1, Paper, Permanente, a base di gomma, Thermal transfer, bianco, 360X-Plus II, 76×50 mm, 1000 labels/roll
SFr. 96.10
1 * 1 ROU
Printable, paper labels for thermal transfer printers.
  • Good contrast and smear resistance
Labels are ideally suited for use on smooth surfaces.
  • Stampante:
    360X-Plus II
  • Printing method:
    Thermal transfer
  • Tipo:
    B-424
  • Colore:
    bianco
  • Descrizione:
    Printer labels
  • Dimensioni:
    76×50 mm
  • Materiale:
    Paper
  • Seq:
    05
  • Modello:
    THT-19-424-1
  • Confezione:
    1000 labels/roll
  • Adesivo:
    Permanente, a base di gomma