Labels, hazard warning
Hazard labels: low temperature, PIC 323-TRI 200-B7541, Poliestere, nero on giallo, 200×173 mm, 1 label/card
Codice catalogo BRDY250271
Nome del fornitore: Brady
Vai a
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifiche
- Colore:nero on giallo
- Descrizione:Hazard labels: low temperature
- Dimensioni:200×173 mm
- Materiale:Poliestere
- Seq:42
- Modello:PIC 323-TRI 200-B7541
- Confezione:1 label/card