Orders LinkContactUsLinkComponent
Recombinant streptavidin
Recombinant streptavidin
Streptavidin, recombinant, 25 mg
Codice catalogo 786-585
Nome del fornitore:   G-Biosciences
Recombinant streptavidin
Codice catalogo 786-585
Nome del fornitore:  G-Biosciences
Numero CAS:  
Recombinant streptavidin
Recombinant streptavidin
Streptavidin, recombinant, 25 mg
Numero CAS:  
Price
908.00 SFr.
Log in per disponibilità
1 * 25 mg
Product Details & Documents
Recombinant streptavidin from Streptomyces avidinii, produced in E.coli.
Streptavidin is a tetrameric protein that has no carbohydrate and thereby reduces the amount of non specific binding. The solubility of streptavidin (isoelectric pH 5) in aqueous buffer is lower than avidin, but the binding of streptavidin to biotin is similar to that of avidin.
Documents
Specifiche
Specifiche
  • Conf:
    25 mg
  • Descrizione:
    Streptavidin, recombinant