Orders LinkContactUsLinkComponent
Blue Dextran loading dye
Blue Dextran loading dye
10 mL
Codice catalogo 786-574
Nome del fornitore:   G-Biosciences
Blue Dextran loading dye
Codice catalogo 786-574
Nome del fornitore:  G-Biosciences
Numero CAS:  87915-38-6
Blue Dextran loading dye
Blue Dextran loading dye
10 mL
Numero CAS:  87915-38-6
Price
SFr. 175.00
Log in per disponibilità
1 * 10 mL
Product Details & Documents
A proprietary mixture of Formaldehyde, EDTA and Blue Dextran that is designed for use in loading DNA samples onto denaturing polyacrylamide gels.
Commonly used in loading DNA samples onto denaturing polyacrylamide gels.
Sinonimi:  Dextran, blue
  • Numero CAS:87915-38-6
  • Formula:(C₂₉H₂₁N₇O₁₂S₃·3Na)n
  • Numero MDL:MFCD00130661
Specifiche
Specifiche
  • Conf:
    10 mL