Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
XCHANGE® syringes

XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for LC, 22, 1000 µl, 56 mm, 0,71 mm, LC
Catalog # 548-1818
XCHANGE® syringes
Catalog # 548-1818
XCHANGE® syringes
XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for LC, 22, 1000 µl, 56 mm, 0,71 mm, LC

Price
SFr. 132.00
Log in for availability
1 * 1 ST
Product Details & Documents

The XCHANGE® coupling system secures and locks the syringe in place. This is achieved by a coupling mechanical piece that captures the syringe and a magnet that automatically aligns – eliminating the need for springs, screws or alignment gauges. Once engaged the syringe is locked into position requiring zero holding force.

  • Injection of small and large volumes in one sample sequence
  • Adding of internal standards without cross-contamination
  • Preventative exchange of syringe when using harmful media
Specifications
  • Ø est:
    0,71 mm
  • Gauge:
    22
  • Tipo punta:
    LC
  • Volume:
    1000 µl
  • Descrizione:
    XCHANGE® CTC PAL-XT syringe for LC
  • Lunghezza:
    56 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.