Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
XCHANGE® syringes

XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for GC, 23, 1000 µl, 56 mm, 0,63 mm, Bevelled
Codice catalogo 548-1817
Nome del fornitore:   Trajan Scientific and Medical
XCHANGE® syringes
Codice catalogo 548-1817
Nome del fornitore:  Trajan Scientific and Medical
XCHANGE® syringes
XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for GC, 23, 1000 µl, 56 mm, 0,63 mm, Bevelled

Price
175.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

The XCHANGE® coupling system secures and locks the syringe in place. This is achieved by a coupling mechanical piece that captures the syringe and a magnet that automatically aligns – eliminating the need for springs, screws or alignment gauges. Once engaged the syringe is locked into position requiring zero holding force.

  • Injection of small and large volumes in one sample sequence
  • Adding of internal standards without cross-contamination
  • Preventative exchange of syringe when using harmful media
Specifiche
  • Ø est:
    0,63 mm
  • Gauge:
    23
  • Tipo punta:
    Bevelled
  • Volume:
    1000 µl
  • Descrizione:
    XCHANGE® CTC PAL-XT syringe for GC
  • Lunghezza:
    56 mm
  • Seq:
    08