Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for GC, 23, 25 µl, 56 mm, 0,63 mm, Cone
Codice catalogo 548-1807
Nome del fornitore:   Trajan Scientific and Medical
XCHANGE® syringes
Codice catalogo 548-1807
Nome del fornitore:  Trajan Scientific and Medical
XCHANGE® syringes
XCHANGE® syringes

XCHANGE® CTC PAL-XT syringe for GC, 23, 25 µl, 56 mm, 0,63 mm, Cone

Price
124.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

The XCHANGE® coupling system secures and locks the syringe in place. This is achieved by a coupling mechanical piece that captures the syringe and a magnet that automatically aligns – eliminating the need for springs, screws or alignment gauges. Once engaged the syringe is locked into position requiring zero holding force.

  • Injection of small and large volumes in one sample sequence
  • Adding of internal standards without cross-contamination
  • Preventative exchange of syringe when using harmful media
Specifiche
  • Ø est:
    0,63 mm
  • Gauge:
    23
  • Tipo punta:
    Cone
  • Volume:
    25 µl
  • Descrizione:
    XCHANGE® CTC PAL-XT syringe for GC
  • Lunghezza:
    56 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.