PIG® encapsulating and neutralizing absorbents
Acid encapsulating and neutralising socks, PP with sodium carbonate/alizarin filler, 5,7 L, 4 socks/box, 50×1220 mm
Codice catalogo NEWPPIG309
Nome del fornitore: NEW PIG
PIG® encapsulating and neutralizing absorbents
Codice catalogo NEWPPIG309
Nome del fornitore: NEW PIG
Vai a
Product Details & Documents
One easy step for safer acid or caustic cleanup.
- Choose from mats and socks to help encapsulate and neutralise common acids or caustics in one step
- Colour changes as liquids are taken in, then remains in a colour-changed state when neutralisation is complete; helps identify the best time to handle with the least amount of risk
- Instantly absorbs up to 98% concentrations of sulfuric acid
- Gelling agents solidify acids during neutralisation; prevents them from leaching out of sock
Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.
Specifiche
- Descrizione:Acid encapsulating and neutralising socks, PP with sodium carbonate/alizarin filler
- Assorbanza:5,7 L
- Confezione:4 socks/box
- Ø×L:50×1220 mm



