Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Thermal transfer ribbon for BBP11 printer

Thermal transfer ribbon for BBP11 printer

Diesel resistant ribbon for thermal printing, width 102 mm, BBP11, 74 m, nero
Catalog # BRDY622115
Supplier:   Brady
Thermal transfer ribbon for BBP11 printer
Catalog # BRDY622115
Supplier:  Brady
Thermal transfer ribbon for BBP11 printer
Thermal transfer ribbon for BBP11 printer

Diesel resistant ribbon for thermal printing, width 102 mm, BBP11, 74 m, nero

Price
SFr. 83.00
Log in for availability
1 * 1 ST
Product Details & Documents
  • Strong environmental resistance against smearing and chemicals
  • Reliable, sharp-edge print quality to maximise bar code readability
Specifications
  • Colore:
    nero
  • Descrizione:
    Diesel resistant ribbon for thermal printing, width 102 mm
  • Lunghezza:
    74 m
  • Seq:
    02
  • Per:
    BBP11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.