Reusable earplugs, E-A-R™ Express™
Product Details & Documents
E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can reduce noise levels by 28 dB, and have a hygienic insertion grip.
- Easy to slide gently into ears with slow recovery foam
- Foam tip gives a comfortable and effective seal
- Features a hygienic insertion grip
- Recognisable blue stem and yellow tip for easy identification
Protect hearing at work or during leisure activities with the E-A-R™ Express™ Reusable Earplugs. Designed to slide gently into the ear, the specially designed pod foam tip provides a comfortable and effective seal. The insertion grip eliminates the need to touch the foam, making them extremely hygienic. Get the best of both worlds with a patented slow recovery foam for a comfortable acoustic seal with quick and easy push-to-fit insertion.
Available in a recognisable yellow and blue for easy identification. Provides noise reduction to 28 dB SNR and fully tested and approved against EN 352-2. Model is compatible for use with the e 3M™ E-A-RFit™ Dual-Ear Validation System and available as a probed plug.
Certifications: EN 352-2
Attenzione: * One size fits all.
Specifiche
- Colore:giallo/blu
- Descrizione:Polyurethane earplug, without cord
- Seq:02
- Taglia:Universal