Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
PIG® encapsulating and neutralizing absorbents

PIG® encapsulating and neutralizing absorbents

Acid encapsulating and neutralising mats, PP with sodium carbonate/alizarin, 2,4 L, 25 pads/box, 380×280 mm
Codice catalogo 121-0052
Nome del fornitore:   NEW PIG
PIG® encapsulating and neutralizing absorbents
Codice catalogo 121-0052
Nome del fornitore:  NEW PIG
PIG® encapsulating and neutralizing absorbents
PIG® encapsulating and neutralizing absorbents

Acid encapsulating and neutralising mats, PP with sodium carbonate/alizarin, 2,4 L, 25 pads/box, 380×280 mm

Price
SFr. 133.00
Log in per disponibilità
1 * 25 ST
Product Details & Documents

One easy step for safer acid or caustic cleanup.

  • Choose from mats and socks to help encapsulate and neutralise common acids or caustics in one step
  • Colour changes as liquids are taken in, then remains in a colour-changed state when neutralisation is complete; helps identify the best time to handle with the least amount of risk
  • Instantly absorbs up to 98% concentrations of sulfuric acid
  • Gelling agents solidify acids during neutralisation; prevents them from leaching out of sock

Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.

Specifiche
  • Descrizione:
    Acid encapsulating and neutralising mats, PP with sodium carbonate/alizarin
  • L×L:
    380×280 mm
  • Assorbanza:
    2,4 L
  • Confezione:
    25 pads/box
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..