Accessories for comparators
Manganese, 3/169, 0-4,0 mg/l
Codice catalogo LOVI230690
Nome del fornitore: Lovibond
Vai a
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifiche
- Range di misura:0-4,0 mg/l
- Seq:38
- Codice:3/169
- Metodo di prova:Manganese