Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Heating tapes, HBRT

Self-limiting heating tape with high working temperature, width 11 mm, 54 W
Codice catalogo HORS021500
Nome del fornitore:   HORST
Heating tapes, HBRT
Codice catalogo HORS021500
Nome del fornitore:  HORST
Heating tapes, HBRT

Self-limiting heating tape with high working temperature, width 11 mm, 54 W

Price
Richiedi prezzo
Log in per disponibilità
1 * 1 m
Product Details & Documents

These self-limiting heating tape comes with an additional protective braid and humidity proof. It can be used for installations and are cleaned with steam.

  • Nominal temperature 1200 °C
  • With minimum bending radius of 20 mm
  • No starting current required when the heating circuit starts working
Specifiche
  • Descrizione:
    Self-limiting heating tape with high working temperature, width 11 mm, 54 W
  • Seq:
    01