Adhesive seals
EASYseal adhesive film, non sterile, 80×140 mm, Transparent, -
Codice catalogo GREI676001BE
Vai a
Discontinued
Il prodotto è statodiscontinuato. Tuttavia abbiamo trovato prodotti alternativi simili.
Alternative
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
- Free from RNase, DNase and human DNA
- Non pyrogenic
Specifiche
- Proprietà ottica:Transparent
- Sterili:-
- Descrizione:EASYseal adhesive film, non sterile, 80×140 mm
- Seq:03
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.