Orders LinkContactUsLinkComponent
Height adjustable worktable | Avantor
Height adjustable worktable
Concept hand crank table (height adjustment with hand crank), 1500× 750× 675/1025 mm
Katalognr. SOVEC10141037
Anbieter:   TRESTON
Height adjustable worktable
Katalognr. SOVEC10141037
Anbieter:  TRESTON
Height adjustable worktable
Height adjustable worktable
Concept hand crank table (height adjustment with hand crank), 1500× 750× 675/1025 mm
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These worktables are a suitable solution for industrial laboratories and research and development centers. They are designed to accommodate the necessary measurement equipment and other tools ergonomically at a safe distance for use. These laboratory benches fit also to education purposes. Modularity and accessories are individually adjustable according the task and human requirements.
  • Robust, elegant, and very flexible
  • Easy to adapt to individual needs, new equipment, and changes in the working environment
  • With durable table frame
  • Leveling glides
Bestellinformationen: Table top sold separately; contact your Avantor representative for more information.
Technische Daten
  • Descrizione:
    Concept hand crank table (height adjustment with hand crank)
  • L×P×A:
    1500× 750× 675/1025 mm
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..