Height adjustable worktable
Concept table (stepless height adjustment), 1500× 750× 670/1120 mm
Catalog # SOVEC10041037
Supplier: TRESTON
Jump to:
Product Details & Documents
These worktables are a suitable solution for industrial laboratories and research and development centers. They are designed to accommodate the necessary measurement equipment and other tools ergonomically at a safe distance for use. These laboratory benches fit also to education purposes. Modularity and accessories are individually adjustable according the task and human requirements.
- Robust, elegant, and very flexible
- Easy to adapt to individual needs, new equipment, and changes in the working environment
- With durable table frame
- Leveling glides
Ordering information: Table top sold separately; contact your Avantor representative for more information.
Specifications
- Descrizione:Concept table (stepless height adjustment)
- L×P×A:1500× 750× 670/1120 mm