Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

100 ml, 0,080 ± ml, giallo
Codice catalogo KAVA632433119030
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Codice catalogo KAVA632433119030
Nome del fornitore:  
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

100 ml, 0,080 ± ml, giallo

Discontinued
Product Details & Documents

Soda-lime glass.

  • Enlarged form
  • Non autoclavable at 121 °C

Certifications: DIN 12691, ISO 648

Specifiche
  • Codice colore:
    giallo
  • Capacità:
    100 ml
  • Seq:
    13
  • Tolleranza:
    0,080 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.