Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

1 ml, 0,008 ± ml, blu
  KAVAN632433111014
 :   Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
  KAVAN632433111014
 :  Kavalier
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS
Bulb pipettes, Qualicolor, colour-coded, one mark, Class AS

1 ml, 0,008 ± ml, blu

 
SFr. 671.00
   
1 * 50 ST
 

Soda-lime glass.

  • Enlarged form
  • Non autoclavable at 121 °C

Certifications: DIN 12691, ISO 648

 
  • Codice colore:
    blu
  • Capacità:
    1 ml
  • Seq:
    01
  • Tolleranza:
    0,008 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.