Filling cannulae
Filling cannulae, G 20, 150 mm
Codice catalogo MURRM720/08
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
Ordering information: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Datasheet
Specifiche
- Gauge:G 20
- Descrizione:Filling cannulae
- Lunghezza:150 mm
- Seq:020