Filling cannulae
Filling cannulae, G 12, 76 mm
Catalog # MURRM712/05
Supplier: MURRAY S
Jump to:
Product Details & Documents
Filling cannulae with blunt end and Luer lock fitting.
: Different sizes and gauges are available.
Documents
MURRM714^06_TDS_EN
Scheda dati
Specifications
- Gauge:G 12
- Descrizione:Filling cannulae
- Lunghezza:76 mm
- Seq:006