Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Transport container with screw cap

Transport container with screw cap

40 ml, 30 mm, 85 mm, with insert/print, 500/500 (bag/ctn)
Catalog # BOET08.013.0600
Supplier:   BOETTGER PAUL
Transport container with screw cap
Catalog # BOET08.013.0600
Supplier:  BOETTGER PAUL
Transport container with screw cap
Transport container with screw cap

40 ml, 30 mm, 85 mm, with insert/print, 500/500 (bag/ctn)

Price
SFr. 412.00
Log in for availability
1 * 500 ST
Product Details & Documents

Made of PP, translucent. Mailing containers for medical and biohazard samples.

  • Optimal protection from external elements through cylindrical design
  • Absorbent paper insert provides additional protection, easy to change, multiple-use very simple
  • Biohazard and recycling symbol clearly visible

Mailing containers Ø 30 mm fits into current mailboxes with 35 mm-wide slot, containers 44/114 mm suitable for mailing common culture mediums and containers 30/126 mm with cap suitable for Port-a-Cul.

Certifications: Corresponding to DIN EN 829 (with insert/print).

Ordering information: Transport tubes and screw caps have to be ordered separate.

Caution: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904

Specifications
  • Ø est:
    30 mm
  • Confezione:
    500/500 (bag/ctn)
  • Versione:
    with insert/print
  • Capacità:
    40 ml
  • Altezza:
    85 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.