Sampling valves
ESP valve, with 1/4 inches NPTM HALAR
Codice catalogo MILXYY20040H0
Nome del fornitore: Merck
Vai a
Product Details & Documents
The sampling valves are used for sterile sampling of water or other final products in the pharmaceutical industry.
- Collect samples mid-stream and filter through a MicropreSure unit, or sample into sterile sampling bags or a graduated cylinder
- Samples are kept in sanitary condition for immediate sampling with alcohol or hydrogen peroxide injections
Materials: stem and body: 316L stainless steel or ECTFE body in ECTFE-type units, valve cap: ABS or PP, O-ring and plug seal: silicone or fluoroelastomer.
Certifications: Meets the FDA indirect food requirements cited in code of federal regulations CFR 21, part 177-182.
Specifiche
- Descrizione:ESP valve, with 1/4 inches NPTM HALAR