General purpose sample storage vials and caps
22,00 ml, 23×85 mm, trasparente, Yes, 20-400, PP, PTFE/silicone
Catalog # HEWL5183-4313
Supplier: AGILENT HEWLETT PACKARD
General purpose sample storage vials and caps
Catalog # HEWL5183-4313
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Tappo incluso:Yes
- Materiale tappo:PP
- Dimensione tappo:20-400
- Ø×A:23×85 mm
- Setto:PTFE/silicone
- Capacità:22,00 ml
- Colore:trasparente
- Seq:06