Reference electrode filling solutions
FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml, 6
Nome del fornitore: Mettler - Toledo
Reference electrode filling solutions
Codice catalogo METT51350084
Nome del fornitore: Mettler - Toledo
Numero CAS:
Vai a
Product Details & Documents
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.
Documents
Specifiche
Specifiche
- Conf:6
- Descrizione:FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml