Orders LinkContactUsLinkComponent
Maintenance solutions for electrodes
Maintenance solutions for electrodes
Reactivation solution for regeneration of glass electrodes, treatment time about 1 min., 25 ml, 25 mL
Codice catalogo METT51350104
Nome del fornitore:   Mettler - Toledo
Maintenance solutions for electrodes
Codice catalogo METT51350104
Nome del fornitore:  Mettler - Toledo
Numero CAS:  
Maintenance solutions for electrodes
Reactivation solution for regeneration of glass electrodes, treatment time about 1 min., 25 ml, 25 mL
Numero CAS:  
Price
33.10 SFr.
Log in per disponibilità
1 * 25 mL
Product Details & Documents
Specifiche
Specifiche
  • Conf:
    25 mL
  • Descrizione:
    Reactivation solution for regeneration of glass electrodes, treatment time about 1 min., 25 ml