Orders LinkContactUsLinkComponent
GC capillary column tools
Capillary ceramic tube cutter
Codice catalogo 568-0470
Nome del fornitore:   Trajan Scientific and Medical
GC capillary column tools
Codice catalogo 568-0470
Nome del fornitore:  Trajan Scientific and Medical
GC capillary column tools
Capillary ceramic tube cutter
Price
28.70 SFr.
Log in per disponibilità
1 * 3 ST
Product Details & Documents
The capillary cutting tool is used for burr free cutting of fused silica tubing.
  • Reduces likelihood of blockages
  • Ensures square ends for 'Press fit' style connectors
  • Clean entry for on-column injection
Specifiche
  • Descrizione:
    Capillary ceramic tube cutter
  • Seq:
    04