Adapters, sperical joints
Socket S 51 with steel thread M 48×1,5, O.Ø 36×2,4 mm, I.Ø 31,2 mm
Catalog # JUCH20457.51
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
For reaction vessels, 316 stainless steel, thread length 14 mm.
Specifications
- Tipo:Socket S 51 with steel thread M 48×1,5, O.Ø 36×2,4 mm, I.Ø 31,2 mm