Retort rings
Ring with bosshead, 290 g, 160 mm
Katalognr. JUCHVL04980
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Technische Daten
- Ø est:160 mm
- Descrizione:Ring with bosshead, 290 g
- Seq:11