Retort rings
Ring without bosshead, 107 g, 50 mm
Catalog # JUCHVL04890
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Specifications
- Ø est:50 mm
- Descrizione:Ring without bosshead, 107 g
- Seq:01