Orders LinkContactUsLinkComponent
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da., 1 g, 1
Codice catalogo VARIPL2022-5001
Nome del fornitore:   VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Codice catalogo VARIPL2022-5001
Nome del fornitore:  VARIAN
Numero CAS:  
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da., 1 g, 1
Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
  • Many solvent options increase applicability
  • Stringent quality control improves performance
  • Proprietary manufacturing methods ensure consistent supply
Specifiche
Specifiche
  • Conf:
    1
  • Descrizione:
    Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,08, nominal Mp 2,000 Da.
  • Pk Info:
    1 g
Specifiche