Polymethylmethacrylate standards, GPC/SEC columns calibration
Calibration kit, M-L-10, 10×0,5 g, nominal Mp 600-50000 g/mol, 1
Codice catalogo VARIPL2020-0100
Nome del fornitore: VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Codice catalogo VARIPL2020-0100
Nome del fornitore: VARIAN
Numero CAS:
Vai a
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
- Many solvent options increase applicability
- Stringent quality control improves performance
- Proprietary manufacturing methods ensure consistent supply
Specifiche
Specifiche
- Conf:1
- Descrizione:Calibration kit, M-L-10, 10×0,5 g, nominal Mp 600-50000 g/mol