Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g, 1
Codice catalogo VARIPL2013-3010
Nome del fornitore: VARIAN
Polystyrene standards, GPC/SEC columns calibration
Codice catalogo VARIPL2013-3010
Nome del fornitore: VARIAN
Numero CAS:
Vai a
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Conf:1
- Descrizione:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,03, quantities 10 g