Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-1, 580 - 7500000 g/mol, 1
Codice catalogo VARIPL2010-0501
Nome del fornitore: VARIAN
Pre-prepared polystyrene kits, EasiCal™
Codice catalogo VARIPL2010-0501
Nome del fornitore: VARIAN
Numero CAS:
Vai a
Product Details & Documents
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
- Easy three step process with no fuss
- Cost effective
- Only two injections for improved productivity
- Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.
Specifiche
Specifiche
- Conf:1
- Descrizione:Polystyrene PS-1, 580 - 7500000 g/mol