Orders LinkContactUsLinkComponent
Pre-weighted calibration kits, EasiVial™
PS-H Tri-Pack, 2 ml, 1
  VARIPL2010-0202
 :   VARIAN
Pre-weighted calibration kits, EasiVial™
  VARIPL2010-0202
 :  VARIAN
 :  
Pre-weighted calibration kits, EasiVial™
PS-H Tri-Pack, 2 ml, 1
 :  
1 * 1 ST
The quickest and most convenient method to deliver an accurate 12 point column calibration for organic and aqueous GPC/SEC column calibration.
  • Pre-prepared, time saving product for rapid and reliable GPC column calibration
  • Eliminates tedious weighing procedures to improve calibration accuracy
  • Reduces solvent dispensing to limit risks associated with handling solvents
  • For conventional and multi detector GPC to maximize applicability
The mass of each polymer in the vial is accurately known, so that upon addition of a fixed volume of eluent, the solution is prepared at a precise concentration. It is ideal for both conventional and multi detector GPC calibration. Simple to prepare and manually inject or transfer to autosampler vials or place directly into a compatible autosampler.
  • Conf:
    1
  • Descrizione:
    PS-H Tri-Pack, 2 ml