Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 40, 8 µm, 7,50 mm, 300 mm
  VARIPL1149-6840
 :   VARIAN
Columns and standards for GPC/SEC
  VARIPL1149-6840
 :  VARIAN
Columns and standards for GPC/SEC
Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 40, 8 µm, 7,50 mm, 300 mm

 
 
   
1 * 1 ST
 
 
  • Ø int.:
    7,50 mm
  • Dimensioni particelle:
    8 µm
  • Descrizione:
    Aqueous SEC of polymers, PL-aquagel-OH 40
  • Lunghezza:
    300 mm