Capillary columns, select permanent gases
Columns, select permanent gases/HR, with restek Y-splitters, temp. limits 300/325 °C, Capillare, Select Permanent Gases
Codice catalogo VARICP914310
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
Select Permanent Gases/CO₂ is a set of two parallel columns that combines CP-Molsieve 5 Å for permanent gas analysis and CP-PoraBOND Q for CO₂ analysis.
- Temperature stability up to 300 °C allows short regeneration times and improves efficiency
- One injector, one detector simplifies operation
- Engineered for fast separation, low level analysis and quantification of argon/oxygen
- Separates permanent gases and CO₂ in a single run
Column dimensions meet the need for fast separation, lowest level analysis, and quantification of argon/oxygen. The Select Permanent Gases column separates permanent gases and CO₂ in a single run, and columns are coupled, tested, and securely mounted on the EZ-GRIP column mounting system.
Specifiche
- Formato della colonna:Capillare
- Descrizione:Columns, select permanent gases/HR, with restek Y-splitters, temp. limits 300/325 °C
- Fase:Select Permanent Gases