Orders LinkContactUsLinkComponent
Capillary columns, select permanent gases
Capillary columns, select permanent gases
Columns, select permanent gases/HR, with restek Y-splitters, temp. limits 300/325 °C, Capillare, Select Permanent Gases
Codice catalogo VARICP914310
Nome del fornitore:   VARIAN
Capillary columns, select permanent gases
Codice catalogo VARICP914310
Nome del fornitore:  VARIAN
Capillary columns, select permanent gases
Capillary columns, select permanent gases
Columns, select permanent gases/HR, with restek Y-splitters, temp. limits 300/325 °C, Capillare, Select Permanent Gases
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Select Permanent Gases/CO₂ is a set of two parallel columns that combines CP-Molsieve 5 Å for permanent gas analysis and CP-PoraBOND Q for CO₂ analysis.
  • Temperature stability up to 300 °C allows short regeneration times and improves efficiency
  • One injector, one detector simplifies operation
  • Engineered for fast separation, low level analysis and quantification of argon/oxygen
  • Separates permanent gases and CO₂ in a single run
Column dimensions meet the need for fast separation, lowest level analysis, and quantification of argon/oxygen. The Select Permanent Gases column separates permanent gases and CO₂ in a single run, and columns are coupled, tested, and securely mounted on the EZ-GRIP column mounting system.
Specifiche
  • Formato della colonna:
    Capillare
  • Descrizione:
    Columns, select permanent gases/HR, with restek Y-splitters, temp. limits 300/325 °C
  • Fase:
    Select Permanent Gases