HPLC syringes
Autosampler syringe, -, 500 µl, - mm, AS1000, AS3000
Codice catalogo 555-8239
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
- Easy and accurate
- Reproducible manual injections
Applications include industrial chromatography.
Specifiche
- Per:AS1000, AS3000
- Gauge:-
- Volume:500 µl
- Descrizione:Autosampler syringe
- Lunghezza:- mm
- Seq:08