Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
HPLC syringes

HPLC syringes

Standard fixed needle, 22, 500 µl, 50 mm, 90° blunt end, Rheodyne/Valco™
Codice catalogo 555-8236
Nome del fornitore:   Thermo Fisher Scientific
HPLC syringes
Codice catalogo 555-8236
Nome del fornitore:  Thermo Fisher Scientific
HPLC syringes
HPLC syringes

Standard fixed needle, 22, 500 µl, 50 mm, 90° blunt end, Rheodyne/Valco™

Price
172.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.

  • Easy and accurate
  • Reproducible manual injections

Applications include industrial chromatography.

Specifiche
  • Per:
    Rheodyne/Valco™
  • Gauge:
    22
  • Tipo punta:
    90° blunt end
  • Volume:
    500 µl
  • Descrizione:
    Standard fixed needle
  • Lunghezza:
    50 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.