Orders LinkContactUsLinkComponent
Inlet base seals
Gold-plated inlet base seals, 0,8 mm ID, for use with single column installation
Codice catalogo 555-9038
Nome del fornitore:   Thermo Fisher Scientific
Inlet base seals
Codice catalogo 555-9038
Nome del fornitore:  Thermo Fisher Scientific
Inlet base seals
Inlet base seals
Gold-plated inlet base seals, 0,8 mm ID, for use with single column installation
Price
609.00 SFr.
Log in per disponibilità
1 * 10 ST
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifiche
  • Descrizione:
    Gold-plated inlet base seals, 0,8 mm ID, for use with single column installation
  • Seq:
    01