Inlet base seals
Gold-plated inlet base seals, 1,2 mm ID, for use with dual column installation
Codice catalogo 555-9215
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifiche
- Descrizione:Gold-plated inlet base seals, 1,2 mm ID, for use with dual column installation
- Seq:04