Inlet base seals
Siltek®-treated inlet base seals, 0,8 mm ID
Codice catalogo 555-9222
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifiche
- Descrizione:Siltek®-treated inlet base seals, 0,8 mm ID
- Seq:07