Orders LinkContactUsLinkComponent
UHPLC filter
UHPLC filter
UHPLC direct connect filter holder
Codice catalogo 555-6747
Nome del fornitore:   Thermo Fisher Scientific
UHPLC filter
Codice catalogo 555-6747
Nome del fornitore:  Thermo Fisher Scientific
UHPLC filter
UHPLC filter
UHPLC direct connect filter holder
Price
SFr. 494.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Column protection for Hypersil GOLD™ 1,9 µm and Syncronis™ 1,7 µm columns without compromising performance.
  • Low volume filter cartridge design
  • Maintain peak shape
  • Minimal efficiency loss through dispersion
Specifiche
  • Descrizione:
    UHPLC direct connect filter holder
  • Seq:
    01