UHPLC filter
UHPLC direct connect filter holder
Codice catalogo 555-6747
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Column protection for Hypersil GOLD™ 1,9 µm and Syncronis™ 1,7 µm columns without compromising performance.
- Low volume filter cartridge design
- Maintain peak shape
- Minimal efficiency loss through dispersion
Specifiche
- Descrizione:UHPLC direct connect filter holder
- Seq:01