Caps
Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter, GL 45, 1 port for 1/16" tubing, 6 ports for 1/8" and 1/16" OD tubing, PP
Codice catalogo JOURJR-S-11006
Nome del fornitore: VICI JOUR RESEARCH
Vai a
Product Details & Documents
These caps top bottles are available with two, three, or four colour-coded 1/4-28 thread for ¹/₈ or ¹/₁₆" O.Ø tubing.
Specifiche
- Filettatura:GL 45
- Descrizione:Cap, including EPDM O-ring, nuts, ferrules and colour-sleeve adapter
- Materiale:PP
- Seq:05
- Connettore:1 port for 1/16" tubing, 6 ports for 1/8" and 1/16" OD tubing