Extender tools
Extender tool, Alluminio, Standard micro knurl
Codice catalogo UPCHP-277
Nome del fornitore: UPCHURCH SCIENTIFIC
Vai a
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifiche
- Descrizione:Extender tool
- Materiale:Alluminio
- Use with head style:Standard micro knurl