Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Laboratory trays

bianco, 171×125×35 mm
  SEMA4061
 :   SEMADENI
Laboratory trays
  SEMA4061
 :  SEMADENI
Laboratory trays
Laboratory trays

bianco, 171×125×35 mm

 
SFr. 52.60
   
1 * 100 ST
 

Trays made of white co-polyester.

  • Stable
  • Temperature resistant from −40 to +220 °C
  • Suitable for use as a sterilisation or microwave tray

With rounded corners and smooth surfaces.

 
  • Colore:
    bianco
  • Seq:
    01
  • L×P×A:
    171×125×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.