Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Jars with screw cap

Jars with screw cap

250 ml, 70 mm, 90 mm
Codice catalogo SEMA5919
Nome del fornitore:   SEMADENI
Jars with screw cap
Codice catalogo SEMA5919
Nome del fornitore:  SEMADENI
Jars with screw cap
Jars with screw cap

250 ml, 70 mm, 90 mm

Price
SFr. 155.00
Log in per disponibilità
1 * 100 ST
Product Details & Documents

Jars, made of PP, with tight screw cap.

Suitable for powders, granules, pastes, ointments, waxes, cosmetics and dermatology products.

Specifiche
  • Ø est:
    70 mm
  • Capacità:
    250 ml
  • Altezza:
    90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.