Jars with overlapping lid
10 mm, 5 mm
Codice catalogo SEMA2112
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifiche
- Ø est:10 mm
- Altezza:5 mm
- Seq:01